Assay Details
| Gene Symbol: LOC4345167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795289.2 | XP_015650775.1 | 4 - 5 | 494 | 93 | |
| GenBank mRNA | AK100935.1 | - | 4 - 5 | 483 | 93 | |
More Information
Set Membership: |
|

LOC4345167
Genomic Map