Assay Details
| Gene Symbol: LOC4345646 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793115.2 | XP_015648601.1 | 1 - 2 | 247 | 67 | |
| GenBank mRNA | AK106170.1 | - | 1 - 2 | 207 | 67 | |
More Information
Set Membership: |
|

LOC4345646
Genomic Map