Assay Details
| Gene Symbol: LOC4346080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792716.2 | XP_015648202.1 | 2 - 3 | 416 | 74 | |
| GenBank mRNA | AK105364.1 | - | 2 - 3 | 413 | 74 | |
More Information
Set Membership: |
|

LOC4346080
Genomic Map