Assay Details
| Gene Symbol: LOC4346116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793616.2 | XP_015649102.1 | 1 - 2 | 1026 | 87 | |
| GenBank mRNA | AK100297.1 | - | 1 - 2 | 851 | 87 | |
More Information
Set Membership: |
|

LOC4346116
Genomic Map