Assay Details
| Gene Symbol: LOC4346116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793616.2 | XP_015649102.1 | 4 - 5 | 1347 | 65 | |
| GenBank mRNA | AK100297.1 | - | 4 - 5 | 1172 | 65 | |
More Information
Set Membership: |
|

LOC4346116
Genomic Map