Assay Details
| Gene Symbol: LOC9266191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793903.2 | XP_015649389.1 | 1 - 2 | 157 | 95 | |
| GenBank mRNA | AK122095.1 | - | 1 - 2 | 79 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015793903.2 transcripts.
Set Membership: |
|

LOC9266191
Genomic Map