Assay Details
| Gene Symbol: LOC4346360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015793072.1 | XP_015648558.1 | 5 - 6 | 834 | 66 | |
| GenBank mRNA | AK100851.1 | - | 5 - 6 | 587 | 66 | |
| BT132419.1 | - | 4 - 5 | 320 | 66 | ||
More Information
Set Membership: |
|

LOC4346360
Genomic Map