Assay Details
| Gene Symbol: LOC4346476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757073.2 | XP_015612559.1 | 10 - 11 | 7231 | 100 | |
| GenBank mRNA | AK109602.1 | - | 9 - 10 | 1945 | 100 | |
More Information
Set Membership: |
|

LOC4346476
Genomic Map