Assay Details
| Gene Symbol: LOC4346476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757073.2 | XP_015612559.1 | 3 - 4 | 6466 | 72 | |
| GenBank mRNA | AK109602.1 | - | 2 - 3 | 1180 | 72 | |
More Information
Set Membership: |
|

LOC4346476
Genomic Map