Assay Details
| Gene Symbol: LOC4347141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757197.2 | XP_015612683.1 | 3 - 4 | 832 | 60 | |
| GenBank mRNA | AK062015.1 | - | 3 - 4 | 703 | 60 | |
| AK103447.1 | - | 3 - 4 | 838 | 60 | ||
More Information
Set Membership: |
|

LOC4347141
Genomic Map