Assay Details
| Gene Symbol: LOC4347279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756679.2 | XP_015612165.1 | 10 - 11 | 1486 | 107 | |
| GenBank mRNA | AK060764.1 | - | 10 - 11 | 1326 | 107 | |
| AK070246.1 | - | 4 - 5 | 335 | 107 | ||
| AK100324.1 | - | 10 - 11 | 1493 | 107 | ||
| AK102736.1 | - | 7 - 8 | 2459 | 107 | ||
More Information
Set Membership: |
|

LOC4347279
Genomic Map