Assay Details
| Gene Symbol: LOC4347279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756679.2 | XP_015612165.1 | 11 - 12 | 1579 | 86 | |
| GenBank mRNA | AK060764.1 | - | 11 - 12 | 1419 | 86 | |
| AK070246.1 | - | 5 - 6 | 428 | 86 | ||
| AK100324.1 | - | 11 - 12 | 1586 | 86 | ||
| AK102736.1 | - | 8 - 9 | 2552 | 86 | ||
More Information
Set Membership: |
|

LOC4347279
Genomic Map