Assay Details
| Gene Symbol: LOC4347279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756679.2 | XP_015612165.1 | 15 - 16 | 1980 | 87 | |
| GenBank mRNA | AK060764.1 | - | 15 - 16 | 1820 | 87 | |
| AK070246.1 | - | 9 - 10 | 829 | 87 | ||
| AK100324.1 | - | 15 - 16 | 1987 | 87 | ||
| AK102736.1 | - | 12 - 13 | 2953 | 87 | ||
More Information
Set Membership: |
|

LOC4347279
Genomic Map