Assay Details
| Gene Symbol: LOC4347279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756679.2 | XP_015612165.1 | 16 - 17 | 2064 | 144 | |
| GenBank mRNA | AK060764.1 | - | 16 - 17 | 1904 | 144 | |
| AK070246.1 | - | 10 - 11 | 913 | 144 | ||
| AK100324.1 | - | 16 - 17 | 2071 | 144 | ||
More Information
Set Membership: |
|

LOC4347279
Genomic Map