Assay Details
| Gene Symbol: LOC4347279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756679.2 | XP_015612165.1 | 4 - 5 | 818 | 92 | |
| GenBank mRNA | AK060764.1 | - | 4 - 5 | 656 | 92 | |
| AK100324.1 | - | 4 - 5 | 825 | 92 | ||
More Information
Set Membership: |
|

LOC4347279
Genomic Map