Assay Details
| Gene Symbol: LOC4347279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756679.2 | XP_015612165.1 | 6 - 7 | 976 | 109 | |
| GenBank mRNA | AK060764.1 | - | 6 - 7 | 816 | 109 | |
| AK100324.1 | - | 6 - 7 | 983 | 109 | ||
| AK102736.1 | - | 4 - 5 | 1799 | 109 | ||
More Information
Set Membership: |
|

LOC4347279
Genomic Map