Assay Details
| Gene Symbol: LOC4347279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756679.2 | XP_015612165.1 | 7 - 8 | 1178 | 81 | |
| GenBank mRNA | AK060764.1 | - | 7 - 8 | 1018 | 81 | |
| AK100324.1 | - | 7 - 8 | 1185 | 81 | ||
| AK102736.1 | - | 5 - 6 | 2001 | 81 | ||
More Information
Set Membership: |
|

LOC4347279
Genomic Map