Assay Details
| Gene Symbol: LOC4347317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756223.2 | XP_015611709.1 | 1 - 2 | 1027 | 79 | |
| GenBank mRNA | AK109479.1 | - | 1 - 2 | 1033 | 79 | |
| AK122120.1 | - | 1 - 2 | 949 | 79 | ||
More Information
Set Membership: |
|

LOC4347317
Genomic Map