Assay Details
| Gene Symbol: LOC4347317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756223.2 | XP_015611709.1 | 2 - 3 | 1328 | 90 | |
| GenBank mRNA | AK122120.1 | - | 2 - 3 | 1250 | 90 | |
More Information
Set Membership: |
|

LOC4347317
Genomic Map