Assay Details
| Gene Symbol: LOC4347450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756256.2 | XP_015611742.1 | 6 - 7 | 1054 | 140 | |
| XM_015756257.2 | XP_015611743.1 | 5 - 6 | 966 | 140 | ||
| GenBank mRNA | AK103057.1 | - | 6 - 7 | 878 | 140 | |
| AK105408.1 | - | 6 - 7 | 818 | 140 | ||
| EU605831.1 | - | 6 - 7 | 674 | 140 | ||
More Information
Set Membership: |
|

LOC4347450
Genomic Map