Assay Details
| Gene Symbol: LOC4347450 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756256.2 | XP_015611742.1 | 8 - 9 | 1309 | 147 | |
| XM_015756257.2 | XP_015611743.1 | 7 - 8 | 1221 | 147 | ||
| GenBank mRNA | AK103057.1 | - | 8 - 9 | 1133 | 147 | |
| AK105408.1 | - | 8 - 9 | 1073 | 147 | ||
| EU605831.1 | - | 8 - 9 | 929 | 147 | ||
More Information
Set Membership: |
|

LOC4347450
Genomic Map