Assay Details
| Gene Symbol: LOC4347635 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757135.2 | XP_015612621.1 | 1 - 2 | 747 | 72 | |
| GenBank mRNA | AK071877.1 | - | 1 - 2 | 738 | 72 | |
More Information
Set Membership: |
|

LOC4347635
Genomic Map