Assay Details
| Gene Symbol: LOC4347810 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756206.2 | XP_015611692.1 | 17 - 18 | 4431 | 148 | |
| XM_015756207.2 | XP_015611693.1 | 17 - 18 | 4428 | 148 | ||
| XM_015756208.2 | XP_015611694.1 | 17 - 18 | 4428 | 148 | ||
| XM_015756209.2 | XP_015611695.1 | 17 - 18 | 4426 | 148 | ||
| GenBank mRNA | AK103018.1 | - | 3 - 4 | 403 | 148 | |
More Information
Set Membership: |
|

LOC4347810
Genomic Map