Assay Details
| Gene Symbol: LOC9272017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759447.2 | XP_015614933.1 | 4 - 5 | 1099 | 76 | |
| XM_015759448.2 | XP_015614934.1 | 4 - 5 | 1112 | 76 | ||
| XM_015759449.2 | XP_015614935.1 | 4 - 5 | 1110 | 76 | ||
| GenBank mRNA | AB096003.2 | - | 4 - 5 | 935 | 76 | |
| AB210109.1 | - | 4 - 5 | 935 | 76 | ||
| AK111418.1 | - | 4 - 5 | 1031 | 76 | ||
More Information
Set Membership: |
|

LOC9272017
Genomic Map