Assay Details
| Gene Symbol: LOC9272017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759447.2 | XP_015614933.1 | 7 - 8 | 1356 | 96 | |
| GenBank mRNA | AB096003.2 | - | 7 - 8 | 1192 | 96 | |
| AB210109.1 | - | 7 - 8 | 1192 | 96 | ||
| AK111418.1 | - | 7 - 8 | 1292 | 96 | ||
More Information
Set Membership: |
|

LOC9272017
Genomic Map