Assay Details
| Gene Symbol: LOC9272017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759447.2 | XP_015614933.1 | 8 - 9 | 1440 | 93 | |
| GenBank mRNA | AB096003.2 | - | 8 - 9 | 1276 | 93 | |
| AB210109.1 | - | 8 - 9 | 1276 | 93 | ||
| AK111418.1 | - | 8 - 9 | 1376 | 93 | ||
More Information
Set Membership: |
|

LOC9272017
Genomic Map