Assay Details
| Gene Symbol: LOC4348446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758395.2 | XP_015613881.1 | 1 | 99 | 80 | |
| GenBank mRNA | EU267981.1 | - | 1 | 95 | 80 | |
More Information
Set Membership: |
|

LOC4348446
Genomic Map