Assay Details
| Gene Symbol: LOC4348571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757989.2 | XP_015613475.1 | 1 - 2 | 135 | 50 | |
| GenBank mRNA | AB100408.1 | - | 1 - 2 | 57 | 50 | |
More Information
Set Membership: |
|

LOC4348571
Genomic Map