Assay Details
| Gene Symbol: LOC4348741 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758636.2 | XP_015614122.1 | 1 - 2 | 1714 | 86 | |
| GenBank mRNA | AK069054.1 | - | 1 - 2 | 1721 | 86 | |
More Information
Set Membership: |
|

LOC4348741
Genomic Map