Assay Details
| Gene Symbol: LOC4348870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758651.2 | XP_015614137.1 | 1 - 2 | 412 | 83 | |
| GenBank mRNA | AK105134.1 | - | 1 - 2 | 386 | 83 | |
More Information
Set Membership: |
|

LOC4348870
Genomic Map