Assay Details
| Gene Symbol: LOC4348870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015758651.2 | XP_015614137.1 | 3 - 4 | 594 | 77 | |
| GenBank mRNA | AK105134.1 | - | 3 - 4 | 568 | 77 | |
More Information
Set Membership: |
|

LOC4348870
Genomic Map