Assay Details
| Gene Symbol: LOC4348872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757474.2 | XP_015612960.1 | 1 - 2 | 460 | 61 | |
| GenBank mRNA | AK073804.1 | - | 1 - 2 | 443 | 61 | |
| DQ229365.1 | - | 1 - 2 | 314 | 61 | ||
More Information
Set Membership: |
|

LOC4348872
Genomic Map