Assay Details
| Gene Symbol: LOC4348894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759420.2 | XP_015614906.1 | 6 - 7 | 781 | 82 | |
| GenBank mRNA | AK060164.1 | - | 6 - 7 | 671 | 82 | |
| AK070445.1 | - | 6 - 7 | 783 | 82 | ||
| AK121743.1 | - | 6 - 7 | 716 | 82 | ||
More Information
Set Membership: |
|

LOC4348894
Genomic Map