Assay Details
| Gene Symbol: LOC9270745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759319.1 | XP_015614805.1 | 16 - 17 | 1704 | 82 | |
| GenBank mRNA | AK102336.1 | - | 16 - 17 | 1711 | 82 | |
More Information
Set Membership: |
|

LOC9270745
Genomic Map