Assay Details
| Gene Symbol: LOC9270745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759319.1 | XP_015614805.1 | 26 - 27 | 2818 | 76 | |
| GenBank mRNA | AK102336.1 | - | 26 - 27 | 2825 | 76 | |
More Information
Set Membership: |
|

LOC9270745
Genomic Map