Assay Details
| Gene Symbol: LOC9270745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759319.1 | XP_015614805.1 | 6 - 7 | 817 | 87 | |
| GenBank mRNA | AK059629.1 | - | 3 - 4 | 339 | 87 | |
| AK102336.1 | - | 6 - 7 | 824 | 87 | ||
More Information
Set Membership: |
|

LOC9270745
Genomic Map