Assay Details
| Gene Symbol: LOC9270745 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759319.1 | XP_015614805.1 | 7 - 8 | 852 | 67 | |
| GenBank mRNA | AK059629.1 | - | 4 - 5 | 374 | 67 | |
| AK102336.1 | - | 7 - 8 | 859 | 67 | ||
More Information
Set Membership: |
|

LOC9270745
Genomic Map