Assay Details
| Gene Symbol: LOC4349004 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759459.1 | XP_015614945.1 | 1 - 2 | 165 | 76 | |
| GenBank mRNA | AK068143.1 | - | 1 - 2 | 138 | 76 | |
More Information
Set Membership: |
|

LOC4349004
Genomic Map