Assay Details
| Gene Symbol: LOC4349006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757481.2 | XP_015612967.1 | 2 - 3 | 457 | 98 | |
| GenBank mRNA | AK108318.1 | - | 2 - 3 | 462 | 98 | |
More Information
Set Membership: |
|

LOC4349006
Genomic Map