Assay Details
| Gene Symbol: LOC4349006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757481.2 | XP_015612967.1 | 3 - 4 | 866 | 110 | |
| GenBank mRNA | AK108318.1 | - | 3 - 4 | 871 | 110 | |
More Information
Set Membership: |
|

LOC4349006
Genomic Map