Assay Details
| Gene Symbol: LOC4349225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759525.2 | XP_015615011.1 | 1 - 2 | 434 | 120 | |
| GenBank mRNA | AK111579.1 | - | 1 - 2 | 340 | 120 | |
More Information
Set Membership: |
|

LOC4349225
Genomic Map