Assay Details
| Gene Symbol: LOC4349225 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759525.2 | XP_015615011.1 | 4 - 5 | 1037 | 78 | |
| GenBank mRNA | AK111579.1 | - | 4 - 5 | 943 | 78 | |
More Information
Set Membership: |
|

LOC4349225
Genomic Map