Assay Details
| Gene Symbol: LOC4349894 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015759605.2 | XP_015615091.1 | 8 - 9 | 1132 | 132 | |
| GenBank mRNA | AK240940.1 | - | 8 - 9 | 1121 | 132 | |
More Information
Set Membership: |
|

LOC4349894
Genomic Map