Assay Details
| Gene Symbol: LOC4351617 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762616.2 | XP_015618102.1 | 6 - 7 | 957 | 71 | |
| GenBank mRNA | AK069406.1 | - | 6 - 7 | 951 | 71 | |
More Information
Set Membership: |
|

LOC4351617
Genomic Map