Assay Details
| Gene Symbol: LOC4351665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015762819.2 | XP_015618305.1 | 7 - 8 | 1260 | 120 | |
| GenBank mRNA | AK072318.1 | - | 7 - 8 | 1243 | 120 | |
More Information
Set Membership: |
|

LOC4351665
Genomic Map