Assay Details
| Gene Symbol: LOC4352284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015763883.2 | XP_015619369.1 | 2 - 3 | 1857 | 73 | |
| GenBank mRNA | AK109540.1 | - | 2 - 3 | 1850 | 73 | |
| AK242914.1 | - | 1841 | 73 | |||
More Information
Set Membership: |
|

LOC4352284
Genomic Map