Assay Details
| Gene Symbol: LOC4352344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765199.2 | XP_015620685.1 | 10 - 11 | 1426 | 64 | |
| GenBank mRNA | AB267729.1 | - | 10 - 11 | 1428 | 64 | |
| AK071160.1 | - | 10 - 11 | 1428 | 64 | ||
More Information
Set Membership: |
|

LOC4352344
Genomic Map