Assay Details
| Gene Symbol: LOC4352344 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765199.2 | XP_015620685.1 | 3 - 4 | 439 | 114 | |
| GenBank mRNA | AB267729.1 | - | 3 - 4 | 441 | 114 | |
| AK071160.1 | - | 3 - 4 | 441 | 114 | ||
More Information
Set Membership: |
|

LOC4352344
Genomic Map