Assay Details
| Gene Symbol: LOC4352558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764738.2 | XP_015620224.1 | 1 - 2 | 263 | 75 | |
| XM_015764739.2 | XP_015620225.1 | 1 - 2 | 263 | 75 | ||
More Information
Set Membership: |
|

LOC4352558
Genomic Map