Assay Details
| Gene Symbol: LOC4352558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764738.2 | XP_015620224.1 | 5 - 6 | 738 | 104 | |
| XM_015764739.2 | XP_015620225.1 | 5 - 6 | 738 | 104 | ||
More Information
Set Membership: |
|

LOC4352558
Genomic Map